Learn More
Advanced Cell Diagnostics RNAscope 2.5 LS Probe - HPV16, Chromogenic detection, FFPE tissues, Single-molecule sensitivity, 1-20 copies per cell
Catalog No. NC1772400
Encompass
RNAscope 2.5 LS Probe - HPV16, Chromogenic detection, FFPE tissues, Single-molecule sensitivity, 1-20 copies per cell
Catalog No. NC1772400 Supplier Advanced Cell Diagnostics Supplier No. 311528
RNAscope 2.5 LS Probe - HPV16, Chromogenic detection, FFPE tissues, Single-molecule sensitivity, 1-20 copies per cell