Learn More
Advanced Cell Diagnostics RNAscope® VS Universal AP Detection Reagents
Catalog No. NC3682383
Encompass
RNAscope® VS Universal AP Detection Reagents is a RNA in situ hybridization (ISH) single-plex, chromogenic -RED assay designed for use with the Discovery Ultra automated IHC/ISH slide staining systems by Ventana Medical Systems. RNAscope® VS Universal AP Detection Reagents includes VS Universal amplification detection reagents. Each reagent kit provides enough reagents to stain ~60 slides. Note: To run VS Universal reagents you will need VS 2.5 probes
Catalog No. NC3682383 Supplier Advanced Cell Diagnostics Supplier No. 323260
Add to Cart
Add to Cart
Request a Quote
RNAscope® VS Universal AP Detection Reagents is a RNA in situ hybridization (ISH) single-plex, chromogenic -RED assay designed for use with the Discovery Ultra automated IHC/ISH slide staining systems by Ventana Medical Systems. RNAscope® VS Universal AP Detection Reagents includes VS Universal amplification detection reagents. Each reagent kit provides enough reagents to stain ~60 slides. Note: To run VS Universal reagents you will need VS 2.5 probes