Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ RNase A

Catalog No. FERR1253 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
0.7 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERR1253 0.7 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERR1253 Supplier Thermo Scientific™ Supplier No. R1253
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

RNase A Solution is a component of the GeneJET Plasmid Miniprep Kit and may be purchased separately.

RNase A Solution is a component of the GeneJET Plasmid Miniprep Kit (Cat. No. K0502/K0503) and may be purchased separately. When using with the GenJet Plasmid Miniprep, add the provided RNase A solution to the Resuspension Solution and mix.

RNase A Solution is a component of the GeneJET Plasmid Miniprep Kit (Cat. No. K0502/K0503) and may be purchased separately. When using with the GenJet Plasmid Miniprep, add the provided RNase A solution to the Resuspension Solution and mix. After addition of RNase A, the Resuspension Solution can be used for 6 months when stored at 4°C.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 10 mg/mL
Format Kit
Enzyme RNase
Product Type RNase A
Quantity 0.7 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.