Learn More
LGC GENOMICS LLC (Lucigen) RNase A, 2 mL @ 5 mg/mLEfficiently degrade unwanted RNA in DNA preparations and other samples.
Catalog No. NC9358522
Encompass
This reagent is designed for use with the MasterPure™ kit protocols only.
This item is not returnable. View return policy
Catalog No. NC9358522 Supplier LGC GENOMICS LLC Supplier No. MRNA092
This reagent is designed for use with the MasterPure™ kit protocols only.