Learn More
AG Scientific Inc RNase A, 250 MG
Supplier:  AG Scientific Inc R2000250MG
Encompass_Preferred
RNase stands for ribonuclease, an RNA digesting enzyme. RNase consists of 124 amino acids in one polypeptide chain with 4 disulphide linkages. Ribonucleases can be divided into endoribonucleases and exoribonucleases. RNases have important roles in RNA degradation and turnover in all organisms. Ribonuclease enzyme can unwind the RNA helix by complexing with single-stranded RNA. RNase A is an endoribonuclease with functions in RNA metabolism and regulation of gene expression. It was found to play roles in diseases such as autoimmune diseases, renal insufficiencies and pancreas disorder.CAS Number: 9001-99-4Chemical Formula: C9H14N4O3Storage Temperature: -20CResearch or further manufacturing use only, not for food or drug use.
Catalog No. 50-223-6968