Learn More
Worthington Biochemical Corporation Ribonuclease A, Purified, 100mg
SDP
Catalog No. NC9216994
Encompass
A lyophilized preparation equivalent to Code: RASE. Contains some aggregates as a result of lyophilization but shows no difference in specific activity. Store at 2-8°C. Protect from moisture. Ribonuclease A has a molecular weight of 13,700 daltons. It operates in an optimum pH range of 7.0-7.5.
Catalog No. NC9216994 Supplier Worthington Biochemical Corporation Supplier No. LS005650
A lyophilized preparation equivalent to Code: RASE. Contains some aggregates as a result of lyophilization but shows no difference in specific activity. Store at 2-8°C. Protect from moisture. Ribonuclease A has a molecular weight of 13,700 daltons. It operates in an optimum pH range of 7.0-7.5.