Learn More
Sigma Aldrich Fine Chemicals Biosciences RNase A from bovine pancre
Catalog No. 502901121
Encompass
Bovine pancreatic RNase A is a member of the RNase A protein superfamily. It is one of the most characterized proteins and is a kidney shaped basic protein. This protein is composed of 124 amino acids. In its native form RNase A exists as a homodimer.
Catalog No. 50-290-1121 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 10109169001
Bovine pancreatic RNase A is a member of the RNase A protein superfamily. It is one of the most characterized proteins and is a kidney shaped basic protein. This protein is composed of 124 amino acids. In its native form RNase A exists as a homodimer.