Learn More
Medchemexpress LLC Rnase a, recombinant protease & dnase free, animal free | 9001-99-4 | 10 KU
SDP
Catalog No. 5000340482
Encompass_Preferred
RNase A (Bovine pancreatic RNase) is an endonuclease used in DNA purification by specifically hydrolyzing cytosine or uracil residues of RNA. It degrades RNA in the RNA/DNA duplex and catalyzes the breakdown of 3',5'-phosphodiester linkages of single-stranded RNA. This recombinant RNase A does not contain protease, DNase, or animal components.
  • Specifically hydrolyzes cytosine or uracil residues of RNA.
  • Degrades RNA in the RNA/DNA duplex.
  • Catalyzes the breakdown of 3',5'-phosphodiester linkages of single-stranded RNA.
  • Does not contain protease or DNase.
  • Animal-free formulation.
Catalog No. 50-003-40482 Supplier Medchemexpress LLC Supplier No. HY129046H10KU
Add to Cart
Add to Cart
RNase A (Bovine pancreatic RNase) is an endonuclease used in DNA purification by specifically hydrolyzing cytosine or uracil residues of RNA. It degrades RNA in the RNA/DNA duplex and catalyzes the breakdown of 3',5'-phosphodiester linkages of single-stranded RNA. This recombinant RNase A does not contain protease, DNase, or animal components.
  • Specifically hydrolyzes cytosine or uracil residues of RNA.
  • Degrades RNA in the RNA/DNA duplex.
  • Catalyzes the breakdown of 3',5'-phosphodiester linkages of single-stranded RNA.
  • Does not contain protease or DNase.
  • Animal-free formulation.