Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PureLink™ RNase A (20 mg/mL)

Catalog No. 12091021
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-091-021 10 mL
12-091-039 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-091-021 Supplier Invitrogen™ Supplier No. 12091021
Only null left
Add to Cart
Edge
Add to Cart

PureLink™ RNase A (ribonuclease A) is a bovine pancreatic ribonuclease that cleaves single-stranded RNA.

  • RNase A (ribonuclease A) is a bovine pancreatic ribonuclease that cleaves single-stranded RNA
  • Used for the isolation of RNA-free DNA

Concentration

  • 20 mg RNase A/mL in 50 mM Tris-HCl (pH 8.0), 10 mM EDTA

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage One 10-mL bottle

Store at 2°C to 8°C.
Concentration 20 mg/mL
Packaging Type Bottle
Product Line UltraPure
Quantity 10 mL
Shipping Condition Room Temperature
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.