Learn More
Bioworld RNase AWAY™ 1L
Catalog No. 50196809
Encompass_Preferred
RNase Away™ removes RNase and DNA contamination from glassware and plastic quickly and economically. Simply apply RNase away to surface or soak as needed and rinse with water. Non-abrasive formulation does not contain strong acids. Effective for use with electrophoresis equipment, glass plates, and other laboratory equipment. Specification: Removes RNase and DNA - Ready to use - Non-abrasive. Molecular Weight : 18.02 g/mol, pH : 6.0 - 8.0 at 25 °C, Solubility : completely miscible, Appearance Form : Liquid, Appearance Color : Clear, Melting Point : 0.0 °C, Boiling Point : 100.0 °C, Density : 1.023 g/cm3. 1L
Catalog No. 50-196-809 Supplier Bioworld Supplier No. 107300021
RNase Away™ removes RNase and DNA contamination from glassware and plastic quickly and economically. Simply apply RNase away to surface or soak as needed and rinse with water. Non-abrasive formulation does not contain strong acids. Effective for use with electrophoresis equipment, glass plates, and other laboratory equipment. Specification: Removes RNase and DNA - Ready to use - Non-abrasive. Molecular Weight : 18.02 g/mol, pH : 6.0 - 8.0 at 25 °C, Solubility : completely miscible, Appearance Form : Liquid, Appearance Color : Clear, Melting Point : 0.0 °C, Boiling Point : 100.0 °C, Density : 1.023 g/cm3. 1L