Learn More
Agilent Technologies RNASE BLOCK
Supplier:  Agilent Technologies 300152
Encompass
RNase Block is a 50-kDa protein that specifically inhibits common eukaryotic RNases including RNase A, RNase B and RNase C. It inactivates RNases by binding noncovalently to the RNase molecule.Only 25 to 40 U is required .
Catalog No. 50-125-523