Learn More
MP Biomedicals™ RNase-Erase Decontaminants
Catalog No. p-83822 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Volume (Metric):
250 mL
Packaging Type:
Spray Bottle
Catalog No. Volume (Metric) Packaging Type
ICN821682 250 mL Spray Bottle
1 options
This item is not returnable. View return policy
Catalog No. ICN821682 Supplier MP Biomedicals™ Supplier No. 04821682
This item is not returnable. View return policy
Completely removes RNase contamination from glass and plastic surfaces
Eliminates the need to bake glassware or use DEPC.
  • Leaves no residues that might inhibit enzymatic reactions
  • Supplied as ready-to-use formula
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 250 mL
Packaging Type Spray Bottle