Learn More
Norgen Biotek Corp RNase-Free DNase I Kit, 200 Reactions
Catalog No. NC0848735
Encompass_Preferred
Norgen's RNA purification kits isolate total RNA with minimal amounts of genomic DNA contamination. However, for some sensitive downstream applications, it may be desirable to remove all traces of residual DNA. Norgen's RNase-free DNAse I Kit, with Enzyme Incubation Buffer, can be used for optional on-column DNase digestion with any of Norgen's RNA purification kits. Alternatively, after isolating total RNA using one of Norgen's RNA purification kits, the RNA elution can be treated with this DNase I. The RNA can then be purified from the DNase using Norgen's RNA Clean-Up and Concentration Kit (Cat# 23600), and the RNA can then be used in downstream applications.
Catalog No. NC0848735 Supplier Norgen Biotek Corp Supplier No. 25720
Norgen's RNA purification kits isolate total RNA with minimal amounts of genomic DNA contamination. However, for some sensitive downstream applications, it may be desirable to remove all traces of residual DNA. Norgen's RNase-free DNAse I Kit, with Enzyme Incubation Buffer, can be used for optional on-column DNase digestion with any of Norgen's RNA purification kits. Alternatively, after isolating total RNA using one of Norgen's RNA purification kits, the RNA elution can be treated with this DNase I. The RNA can then be purified from the DNase using Norgen's RNA Clean-Up and Concentration Kit (Cat# 23600), and the RNA can then be used in downstream applications.