Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biotang Inc RNAse Free Potassium Acetate, 8M, 100 ml

Catalog No. 50152405 Shop All BIOTANG Inc Products
Encompass_Preferred

RNAse Free Potassium Acetate, 8M, 100ml.

Catalog No. 50-152-405 Supplier Biotang Inc Supplier No. BTBB951
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RNAse Free Potassium Acetate, 8M, 100ml.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.