Learn More
Biotang Inc RNASE FREE SSC (20X) 250 ML, R-240, Sodium Citrate 0.3 M, Sodium Chloride 3.0 M, pH 7.0. Prepared in RNase free 18.2 megohms water and filtered through 1 micron filter. Shipped at room temperature. Store at room temperature.
Catalog No. 50152411 Shop All BIOTANG Inc Products
Encompass_Preferred
RNASE FREE SSC (20X) 250 ML, R-240, Sodium Citrate 0.3 M, Sodium Chloride 3.0 M, pH 7.0. Prepared in RNase free 18.2 megohms water and filtered through 1 micron filter. Shipped at room temperature. Store at room temperature.
Catalog No. 50-152-411 Supplier Biotang Inc Supplier No. BTBB957
RNASE FREE SSC (20X) 250 ML, R-240, Sodium Citrate 0.3 M, Sodium Chloride 3.0 M, pH 7.0. Prepared in RNase free 18.2 megohms water and filtered through 1 micron filter. Shipped at room temperature. Store at room temperature.