Learn More
Sigma Aldrich Fine Chemicals Biosciences RNase from bovine pancreas
Catalog No. 502901518
Encompass
Human chromosome 14q11.2 shows the presence of all human RNases. Only eight (RNase 1-8) of them were true called as canonical RNases. And the rest nine to thirteen are called as noncanonical RNases. Human RNase 1 is called as HP-RNase or human pancreatic RNase. Human RNase 2 is also called as EDN (eosinophils derived neurotoxin) that is isolated from eosinophils.
Catalog No. 50-290-1518 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 10109134001
Human chromosome 14q11.2 shows the presence of all human RNases. Only eight (RNase 1-8) of them were true called as canonical RNases. And the rest nine to thirteen are called as noncanonical RNases. Human RNase 1 is called as HP-RNase or human pancreatic RNase. Human RNase 2 is also called as EDN (eosinophils derived neurotoxin) that is isolated from eosinophils.