Learn More
New England Biolabs, Inc. RNase H – 1250 units
SDP
Catalog No. 50811717
Encompass
RNase H (Ribonuclease H ) is an endoribonuclease that specifically hydrolyzes the phosphodiester bonds of RNA which is hybridized to DNA. This enzyme does not digest single or double-stranded DNA.
Catalog No. 50-811-717 Supplier New England Biolabs, Inc. Supplier No. M0297L
RNase H (Ribonuclease H ) is an endoribonuclease that specifically hydrolyzes the phosphodiester bonds of RNA which is hybridized to DNA. This enzyme does not digest single or double-stranded DNA.