Learn More
Cayman Chemical RNase H2BCTRm RabbIt Mon 100uL
SDP
Catalog No. 502913473
Encompass_Preferred
A p300 and CBP inhibitor selectively binds to p300 and CBP over BRD4 (Kds 1 3 1 7 and 222 nM respectively) inhibits proliferation of OPM-2 cells (GI50 5 nM) induces tumor regression in MOLM-16 and OPM-2 mouse xenograft models at 20 mg/kg
Catalog No. 50-291-3473 Supplier Cayman Chemical Supplier No. 35614100 UL
A p300 and CBP inhibitor selectively binds to p300 and CBP over BRD4 (Kds 1 3 1 7 and 222 nM respectively) inhibits proliferation of OPM-2 cells (GI50 5 nM) induces tumor regression in MOLM-16 and OPM-2 mouse xenograft models at 20 mg/kg