Learn More
New England Biolabs, Inc. RNase HII – 250 units
SDP
Catalog No. 50995220
Encompass
Ribonuclease HII (RNase HII) is an endoribonuclease that preferentially nicks 5' to a ribonucleotide within the context of a DNA duplex. The enzyme leaves 5' phosphate and 3' hydroxyl ends. RNase HII will also nick at multiple sites along the RNA portion of an Okazaki fragment.
Catalog No. 50-995-220 Supplier New England Biolabs, Inc. Supplier No. M0288S
Ribonuclease HII (RNase HII) is an endoribonuclease that preferentially nicks 5' to a ribonucleotide within the context of a DNA duplex. The enzyme leaves 5' phosphate and 3' hydroxyl ends. RNase HII will also nick at multiple sites along the RNA portion of an Okazaki fragment.