Learn More
LGC GENOMICS LLC (Lucigen) RNase I, 1000 U @ 10 U/LCompletely digest single-stranded RNA with this heat-labile, non-sequence specific RNase.
Catalog No. NC1304017
Encompass
RNase I degrades single-stranded RNA to nucleoside 3 monophosphates via 2,3 cyclic monophosphate intermediates by cleaving between all dinucleotide pairs,2,3 unlike RNase A, which cleaves only after cytosine and uridine. In addition, the enzyme is completely inactivated by heating at 70C for 15 minutes, eliminating the requirement to remove the enzyme prior to many subsequent procedures.
This item is not returnable. View return policy
Catalog No. NC1304017 Supplier LGC GENOMICS LLC Supplier No. N6901K
RNase I degrades single-stranded RNA to nucleoside 3 monophosphates via 2,3 cyclic monophosphate intermediates by cleaving between all dinucleotide pairs,2,3 unlike RNase A, which cleaves only after cytosine and uridine. In addition, the enzyme is completely inactivated by heating at 70C for 15 minutes, eliminating the requirement to remove the enzyme prior to many subsequent procedures.