Learn More
Apexbio Technology LLC RNase III 1000 U
SDP
Supplier:  Apexbio Technology LLC K30621000
Encompass_Preferred
RNase III is a double-stranded RNA-specific ribonuclease targeting dsRNA substrates It is designed to cleave double-stranded RNA thereby generating short interfering RNA (siRNA) fragments and facilitating gene silencing processes RNase III exerts its biological activity primarily through sequence-specific endonucleolytic cleavage of dsRNA producing siRNA fragments with 3 hydroxyl termini and 2 3 nucleotide overhangs analogous to Dicer-generated products Based on these properties RNase III holds research potential in RNA interference assays gene silencing studies functional genomic target validation and gene-expression regulation experiments
Catalog No. 50-000-96997