Learn More
New England Biolabs, Inc. RNase Inhibitor, Human Placenta – 2000 units
SDP
Catalog No. 50994908
Encompass
RNase Inhibitor, Human Placenta is a recombinant human placental protein which specifically inhibits ribonucleases (RNases) A, B and C. It is not effective against RNase 1, RNase T1, S1 Nuclease, RNase H or RNase from Aspergillus. In addition, no inhibition of polymerase activity is observed when RNase Inhibitor, Human Placenta is used with Taq DNA Polymerase, AMV or M-MuLV Reverse Transcriptases, or Phage RNA Polymerases (SP6, T7, or T3). The 50 kDa protein inhibits RNases by binding noncovalently in a 1:1 ratio with an association constant greater than 1014.
Catalog No. 50-994-908 Supplier New England Biolabs, Inc. Supplier No. M0307S
RNase Inhibitor, Human Placenta is a recombinant human placental protein which specifically inhibits ribonucleases (RNases) A, B and C. It is not effective against RNase 1, RNase T1, S1 Nuclease, RNase H or RNase from Aspergillus. In addition, no inhibition of polymerase activity is observed when RNase Inhibitor, Human Placenta is used with Taq DNA Polymerase, AMV or M-MuLV Reverse Transcriptases, or Phage RNA Polymerases (SP6, T7, or T3). The 50 kDa protein inhibits RNases by binding noncovalently in a 1:1 ratio with an association constant greater than 1014.