Learn More
Macherey-Nagel MACHEREY-NAGEL NucleoSpin RNA Kit, High-Yield RNA Isolation, On-Column rDNase Digest, 250 Preps
Catalog No. NC2164773
Encompass_Preferred
The MACHEREY-NAGEL NucleoSpin RNA Kit is an efficient RNA isolation kit that facilitates the purification of high-quality RNA from various sample types, including cells and tissues. With an included on-column rDNase for DNA removal and quick preparation time, it streamlines workflows for sensitive applications like RT-PCR and RNA-sequencing. Its silica membrane technology ensures reliable results, making it a standout choice for scientists needing effective and dependable RNA purification.
This item is not returnable. View return policy
Catalog No. NC2164773 Supplier Macherey-Nagel Supplier No. 740955.250
The MACHEREY-NAGEL NucleoSpin RNA Kit is an efficient RNA isolation kit that facilitates the purification of high-quality RNA from various sample types, including cells and tissues. With an included on-column rDNase for DNA removal and quick preparation time, it streamlines workflows for sensitive applications like RT-PCR and RNA-sequencing. Its silica membrane technology ensures reliable results, making it a standout choice for scientists needing effective and dependable RNA purification.