Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC RNase L-IN-1 trihydrochloride | 98.1% | 542.67 g/mol | C27H35FN6O3S | 10 MG
SDP

Catalog No. 5000286296
Encompass_Preferred

RNase L-IN-1 trihydrochloride is the trihydrochloride salt form of a small-molecule inhibitor of RNase L, supplied for research use to study RNase L-mediated RNA degradation, antiviral responses, and inflammation. The compound is provided as a solid salt suitable for biochemical and cellular assays.

  • Inhibits RNase L activity for mechanistic and antiviral research.
  • Trihydrochloride salt form improves solubility and handling.
  • High purity (~98.1%) suitable for research applications.
  • Yellow to brown solid appearance, powder form for storage.
  • Molecular formula C27H35FN6O3S; molecular weight 542.67 g/mol.
  • Store powder at -20°C for long-term stability; follow supplier guidelines.

Catalog No. 50-002-86296 Supplier Medchemexpress LLC Supplier No. HY155583A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RNase L-IN-1 trihydrochloride is the trihydrochloride salt form of a small-molecule inhibitor of RNase L, supplied for research use to study RNase L-mediated RNA degradation, antiviral responses, and inflammation. The compound is provided as a solid salt suitable for biochemical and cellular assays.

  • Inhibits RNase L activity for mechanistic and antiviral research.
  • Trihydrochloride salt form improves solubility and handling.
  • High purity (~98.1%) suitable for research applications.
  • Yellow to brown solid appearance, powder form for storage.
  • Molecular formula C27H35FN6O3S; molecular weight 542.67 g/mol.
  • Store powder at -20°C for long-term stability; follow supplier guidelines.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.