Learn More
Bioworld RNase-Off Refill 500ml
Catalog No. 50196897
Encompass_Preferred
Unique formulation destroys RNase & DNase on contact. Completely washed away when rinsed. Spray container prevents waste of reagents and uniformly covers area. Does not contain any toxic agents or leave a residue. Molecular Weight : 18.02 g/mol, pH : 6.0 - 8.0 at 25 °C, Solubility : completely miscible, Appearance Form : Liquid, Appearance Color : Colorless, Melting Point : 0.0 °C, Boiling Point : 100.0 °C, Density : 1.023 g/cm3. 500ml
Catalog No. 50-196-897 Supplier Bioworld Supplier No. 107600461
Unique formulation destroys RNase & DNase on contact. Completely washed away when rinsed. Spray container prevents waste of reagents and uniformly covers area. Does not contain any toxic agents or leave a residue. Molecular Weight : 18.02 g/mol, pH : 6.0 - 8.0 at 25 °C, Solubility : completely miscible, Appearance Form : Liquid, Appearance Color : Colorless, Melting Point : 0.0 °C, Boiling Point : 100.0 °C, Density : 1.023 g/cm3. 500ml