Learn More
Bioworld RNase-Off Refill, 1 L
Catalog No. 50196898
Encompass_Preferred
RNase Off Refill, 500MLUnique formulation destroys RNase & DNase on contact Completely washed away when rinsed Spray container prevents waste of reagents and uniformly covers area Does not contain any toxic agents or leave a residue
Catalog No. 50-196-898 Supplier Bioworld Supplier No. 107600462
RNase Off Refill, 500MLUnique formulation destroys RNase & DNase on contact Completely washed away when rinsed Spray container prevents waste of reagents and uniformly covers area Does not contain any toxic agents or leave a residue