Learn More
Apexbio Technology LLC RNase R (20 U/μL) 2500 U
SDP
Catalog No. 5000097230
Encompass_Preferred
RNase R is a magnesium ion-dependent 3 5 exoribonuclease targeting linear RNA molecules It is designed to selectively digest linear RNAs thereby enriching circular RNA and lariat RNA species in nucleic acid preparations RNase R exerts its biological activity primarily via magnesium-dependent degradation of linear RNA transcripts leaving circular and highly structured RNAs intact Based on these properties RNase R holds research potential in workflows such as circular RNA enrichment RNA sequencing (RNA-seq) gene expression profiling and alternative splicing analyses
Catalog No. 50-000-97230 Supplier Apexbio Technology LLC Supplier No. K30612500
RNase R is a magnesium ion-dependent 3 5 exoribonuclease targeting linear RNA molecules It is designed to selectively digest linear RNAs thereby enriching circular RNA and lariat RNA species in nucleic acid preparations RNase R exerts its biological activity primarily via magnesium-dependent degradation of linear RNA transcripts leaving circular and highly structured RNAs intact Based on these properties RNase R holds research potential in workflows such as circular RNA enrichment RNA sequencing (RNA-seq) gene expression profiling and alternative splicing analyses