Learn More
Abcam RNase R, 1KIT
Catalog No. 502215701
Encompass_Preferred
RNase R is an E. coli exoribonuclease that exhibits 3'-to-5' exonuclease activity, efficiently digesting nearly all linear RNA species. - Efficiently digests linear RNA species with 3'-to-5' exonuclease activity - High purity – contains RNase R at 10 U/µl for optimal performance
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-221-5701 Supplier Abcam Supplier No. AB2869291KIT
RNase R is an E. coli exoribonuclease that exhibits 3'-to-5' exonuclease activity, efficiently digesting nearly all linear RNA species. - Efficiently digests linear RNA species with 3'-to-5' exonuclease activity - High purity – contains RNase R at 10 U/µl for optimal performance
The product is subject to the following: Abcam Restricted Use Statement