Learn More
Apexbio Technology LLC RNase RNA and DNA Remover 500mlx10
SDP
Catalog No. 503107200
Encompass_Preferred
RNase RNA and DNA Remover is a nontoxic spray reagent effectively removing RNase DNA RNA and protein contamination from laboratory benches instruments and consumables creating a clean working environment Regular use of this product is recommended for cleaning benchtops consumables instruments and other solid surfaces related to molecular biology experiments to prevent experimental interference caused by nucleic acid and RNase contamination
Catalog No. 50-310-7200 Supplier Apexbio Technology LLC Supplier No. K11435000
RNase RNA and DNA Remover is a nontoxic spray reagent effectively removing RNase DNA RNA and protein contamination from laboratory benches instruments and consumables creating a clean working environment Regular use of this product is recommended for cleaning benchtops consumables instruments and other solid surfaces related to molecular biology experiments to prevent experimental interference caused by nucleic acid and RNase contamination