Learn More
Meridian Life Science RNase-Tolerant MMLV-RT, 2ML
SDP
Catalog No. 5000856397
Encompass_Preferred
A Moloney Murine Leukemia Virus (MMLV) Reverse Transcriptase (RT), which exhibits high sensitivity and efficiency, and RNase Inhibitor (Ribonuclease Inhibitor) which inhibits a broad spectrum of eukaryotic RNases, including RNases A, B and C. RNase-Tolerant MMLV-RT shows no inhibition of polymerase or reverse transcriptase activity, so can be used in cDNA synthesis or one-step RT-qPCR reactions. RNase-Tolerant MMLV-RT is useful in any applications where the presence of RNases is a potential problem.
Catalog No. 50-008-56397 Supplier Meridian Life Science Supplier No. MDX0432
A Moloney Murine Leukemia Virus (MMLV) Reverse Transcriptase (RT), which exhibits high sensitivity and efficiency, and RNase Inhibitor (Ribonuclease Inhibitor) which inhibits a broad spectrum of eukaryotic RNases, including RNases A, B and C. RNase-Tolerant MMLV-RT shows no inhibition of polymerase or reverse transcriptase activity, so can be used in cDNA synthesis or one-step RT-qPCR reactions. RNase-Tolerant MMLV-RT is useful in any applications where the presence of RNases is a potential problem.