Learn More
Aviva Systems Biology Corp RNASE1 Antibody - middle region (ARP40432_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40432_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RNASE1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RNASE1. Alternate names: MGC12408, RIB1, RNS1ApplicationWB. Species reactivity: Cow, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-53198