Learn More
Novus Biologicals™ RNASE12 Recombinant Protein Antigen

Description
Specifications
Specifications
| Gene ID (Entrez) | 493901 |
| Purity | >80% by SDS-PAGE and Coomassie blue staining |
| Common Name | ribonuclease, RNase A family, 12 (non-active), ribonuclease-like protein 12 |
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Gene Symbol | RNASE12 |
| Molecular Weight (g/mol) | 24 kDa |
| Quantity | 100 μL |
| Source | E. coli |
| Specific Reactivity | This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-54423. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.