Learn More
Aviva Systems Biology Corp RNASE9 Antibody - N-terminal region (ARP48114_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48114_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RNASE9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RNASE9. Alternate names: h461, HEL128ApplicationWB. Species reactivity: Human, Pig
Catalog No. 50-014-59717