Learn More
Aviva Systems Biology Corp RNASEH1 Antibody - middle region (ARP40434_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40434_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RNASEH1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RNASEH1. Alternate names: H1RNA, RNH1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-57450