Learn More
Aviva Systems Biology Corp RNASEH2A Antibody - middle region (ARP40647_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40647_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RNASEH2A. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human RNASEH2A. Alternate names: AGS4, JUNB, RNHL, RNHIA, RNASEHIApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Horse, Human, Mouse, Pig, Rat, Yeast, Zebrafish
Catalog No. 50-014-57570