Learn More
Aviva Systems Biology Corp RNASEH2A Antibody - C-terminal region (ARP40646_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40646_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RNASEH2A. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human RNASEH2A. Alternate names: JUNB, RNASEHI, RNHIA, RNHL, AGS4ApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-57569