Learn More
Aviva Systems Biology Corp RNASEL Antibody - C-terminal region (AVARP09034_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP09034_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RNASEL. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human RNASEL. Alternate names: RNS4, PRCA1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig
Catalog No. 50-014-62579