Learn More
Aviva Systems Biology Corp RNASEN Antibody - middle region (ARP40736_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40736_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RNASEN. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RNASEN. Alternate names: DROSHA, ETOHI2, HSA242976, RANSE3L, RN3, RNASE3L, RNASENApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57604