Learn More
Aviva Systems Biology Corp RNASET2 Antibody - middle region (ARP33127_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33127_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RNASET2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RNASET2. Alternate names: RNASE6PL, bA514O12.3, RP11-514O12.3ApplicationIHC, WB. Species reactivity: Dog, Horse, Human, Rat
Catalog No. 50-014-52892