Learn More
Sigma Aldrich Fine Chemicals Biosciences RNAspin Mini 50 Cytiva 25
Catalog No. 502904854
Encompass
illustra(TM) RNAspin Mini RNA Isolation Kit is designed for rapid extraction of high-quality RNA from a wide range of sample types.illustra(TM) RNAspin Mini Isolation Kit can be used to isolate total RNA from cultured cells tissue bacteria or yeast. The isolated RNA is of a high enough quality and quantity (up to 100 mug) for multiple downstream experiments and sensitive enzymatic applications like RT-PCR primer extension and RNase protection assays. The RNAspin Mini Kit has been used for the isolation of high-quality total RNA from several types of source material including hard-to-lyse bacterial strains and applied in downstream experiments such as quantitative reverse transcriptase PCR (QRT-PCR) Northern blot and microarray experiments. The kit is supplied with a full protocol booklet with a detachable quick reference protocol card and all the necessary components including prefilters and DNase I.We are committed to bringing you Greener Alternative Products which adhere t
Catalog No. 50-290-4854 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE25050071
illustra(TM) RNAspin Mini RNA Isolation Kit is designed for rapid extraction of high-quality RNA from a wide range of sample types.illustra(TM) RNAspin Mini Isolation Kit can be used to isolate total RNA from cultured cells tissue bacteria or yeast. The isolated RNA is of a high enough quality and quantity (up to 100 mug) for multiple downstream experiments and sensitive enzymatic applications like RT-PCR primer extension and RNase protection assays. The RNAspin Mini Kit has been used for the isolation of high-quality total RNA from several types of source material including hard-to-lyse bacterial strains and applied in downstream experiments such as quantitative reverse transcriptase PCR (QRT-PCR) Northern blot and microarray experiments. The kit is supplied with a full protocol booklet with a detachable quick reference protocol card and all the necessary components including prefilters and DNase I.We are committed to bringing you Greener Alternative Products which adhere t