Learn More
Molecular Research Center RNAZOL RT REAGENT 100 ML
SDP
Catalog No. NC9945337
Encompass
A single-step reagent isolating mRNA and small RNA in separate fractions. No chloroform-induced phase separation or refrigerated centrifuge required. No DNase treatment. Higher yields and quality than previous single-step methods.
Catalog No. NC9945337 Supplier Molecular Research Center Supplier No. RN 190100ML
A single-step reagent isolating mRNA and small RNA in separate fractions. No chloroform-induced phase separation or refrigerated centrifuge required. No DNase treatment. Higher yields and quality than previous single-step methods.