Learn More
New Jersey Fluid Systems LLC RND HANDLE NEEDLE VALVE
Catalog No. NC9411452
Encompass
316 Stainless Steel Swagelok Integral Bonnet Needle Valve. Forged Body. 5,000 psi @ 100F; -65F to 450F Regulating Stem. PFA Packing 1/4 in. OD Swagelok Tube Fitting ends.
Catalog No. NC9411452 Supplier New Jersey Fluid Systems LLC Supplier No. SS1RS4
316 Stainless Steel Swagelok Integral Bonnet Needle Valve. Forged Body. 5,000 psi @ 100F; -65F to 450F Regulating Stem. PFA Packing 1/4 in. OD Swagelok Tube Fitting ends.