Learn More
Roboz Surgical Roboz Handle, Round, Knurled, 7.5cm Length, 0.5cm Diameter
SDP
Catalog No. 5000574436
Encompass
This handle is designed for use with miniature scalpel disposable blade assemblies. It features a round, knurled grip for secure handling.
  • Round shape for comfortable gripping
  • Knurled surface ensures a secure hold
  • Length of 7.5 cm
  • Diameter of 0.5 cm
  • For miniature scalpel disposable blade assemblies
Catalog No. 50-005-74436 Supplier Roboz Surgical Supplier No. 374312
This handle is designed for use with miniature scalpel disposable blade assemblies. It features a round, knurled grip for secure handling.
  • Round shape for comfortable gripping
  • Knurled surface ensures a secure hold
  • Length of 7.5 cm
  • Diameter of 0.5 cm
  • For miniature scalpel disposable blade assemblies