Learn More
MSC Destaco Vacuum Pad, Flat, Vertical, 1/4 NPT Female, 1 ea
Catalog No. 5000516212
Encompass
This vertical vacuum pad is designed for industrial material handling applications, featuring a flat profile and a durable nitrile construction. It provides reliable suction and performance for secure object manipulation in various automated or manual processes.
  • Flat pad design for versatile surface contact
  • Durable nitrile material for extended use
  • Integrated 1/4 in. NPT female port for secure connection
  • Vertical mounting configuration
  • Engineered for consistent vacuum hold and release
Catalog No. 50-005-16212 Supplier MSC Supplier No. 88576244
This vertical vacuum pad is designed for industrial material handling applications, featuring a flat profile and a durable nitrile construction. It provides reliable suction and performance for secure object manipulation in various automated or manual processes.
  • Flat pad design for versatile surface contact
  • Durable nitrile material for extended use
  • Integrated 1/4 in. NPT female port for secure connection
  • Vertical mounting configuration
  • Engineered for consistent vacuum hold and release