Learn More
MSC Vikan Bucket, 12 Litre, Brown, Polypropylene, 1 ea
Catalog No. 502815362
Encompass
This award-winning 12-liter brown bucket is designed for transporting cleaning chemicals and both hot and cold ingredients. It features a drip-free spout, a raised hand grip, a sturdy stainless steel handle, and calibrated measuring units, making it suitable for various laboratory and industrial applications.
  • Constructed from 98% polypropylene with an AISI 304 stainless steel handle
  • Features a 12 litre (3.17 gal.) capacity
  • Drip-free spout and raised hand grip for ergonomic handling
  • Calibrated for multiple measuring units (litres, UK and US gal.)
  • Flat side helps prevent spillage during transport
  • Stackable design for efficient storage
  • Complies with FDA Regulation CFR 21 and EU food contact materials regulations
  • Withstands temperatures from -20 °C to 100 °C during usage
Catalog No. 50-281-5362 Supplier MSC Supplier No. 82185109
This award-winning 12-liter brown bucket is designed for transporting cleaning chemicals and both hot and cold ingredients. It features a drip-free spout, a raised hand grip, a sturdy stainless steel handle, and calibrated measuring units, making it suitable for various laboratory and industrial applications.
  • Constructed from 98% polypropylene with an AISI 304 stainless steel handle
  • Features a 12 litre (3.17 gal.) capacity
  • Drip-free spout and raised hand grip for ergonomic handling
  • Calibrated for multiple measuring units (litres, UK and US gal.)
  • Flat side helps prevent spillage during transport
  • Stackable design for efficient storage
  • Complies with FDA Regulation CFR 21 and EU food contact materials regulations
  • Withstands temperatures from -20 °C to 100 °C during usage