Learn More
MSC Rubbermaid Food Handling Container, 32 gal, Resin, Round, NSF 2
Catalog No. 502837199
Encompass
This food handling container is designed for efficient and safe food storage and transport. With a 32 gal capacity, it features durable resin construction and is NSF 2 certified, making it suitable for various commercial and industrial applications.
  • Seamless construction
  • Rounded handles for easy grip
  • Recyclable material
  • Reinforced base for added durability
  • Stackable design for space-saving storage
  • Easy transport and storage
  • NSF 2 certified for food handling
Catalog No. 50-283-7199 Supplier MSC Supplier No. 57855322
This food handling container is designed for efficient and safe food storage and transport. With a 32 gal capacity, it features durable resin construction and is NSF 2 certified, making it suitable for various commercial and industrial applications.
  • Seamless construction
  • Rounded handles for easy grip
  • Recyclable material
  • Reinforced base for added durability
  • Stackable design for space-saving storage
  • Easy transport and storage
  • NSF 2 certified for food handling