Learn More
Cole-Parmer W.S. Tyler 8577 Round Sieve Brush for 120 Mesh Sieves and Finer, Nylon Bristles
Catalog No. NC9940193 Shop All Cole-Parmer Products
Encompass
Use to clean wire cloth test sieves. Round sieve brush is for use with fine mesh sieves, 120 mesh and finer. Brush has nylon bristles and wood handle.
Catalog No. NC9940193 Supplier Cole-Parmer Supplier No. 5998696
Use to clean wire cloth test sieves. Round sieve brush is for use with fine mesh sieves, 120 mesh and finer. Brush has nylon bristles and wood handle.