Learn More
Research Products International Corp Azlon Round Wash Bottles, Ethanol, 500ml, 5/cs
SDP
Catalog No. 502229856
Encompass
Azlon Round Wash Bottles are designed to dispense liquids safely and easily with a fine control precision jet tip. Bottles feature integral inner tube to prevent cross contamination and safe filling. Chemical names are printed in four languages (English, French, German, Spanish), include key safety information, chemical formula, CAS number, GHS symbols, NFPA diamond, signal word for hazard statements and risk phrases. Supplied 5 per case.
Wide mouth
Safety data in four languages
Precision jet tip
Vented cap
Catalog No. 50-222-9856 Supplier Research Products International Corp Supplier No. 249419
Azlon Round Wash Bottles are designed to dispense liquids safely and easily with a fine control precision jet tip. Bottles feature integral inner tube to prevent cross contamination and safe filling. Chemical names are printed in four languages (English, French, German, Spanish), include key safety information, chemical formula, CAS number, GHS symbols, NFPA diamond, signal word for hazard statements and risk phrases. Supplied 5 per case.
Wide mouth
Safety data in four languages
Precision jet tip
Vented cap