Learn More
Aviva Systems Biology Corp RND1 Antibody - C-terminal region (ARP61632_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61632_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RND1. It was validated on Western Blot. . Alternate names: ARHS, FLJ42294, RHO6, RHOSApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-425