Learn More
Aviva Systems Biology Corp RND2 Antibody - C-terminal region (ARP64713_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64713_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RND2. It was validated on Western Blot. . Alternate names: ARHN, RHO7, RhoNApplicationWB. Species reactivity: Human, Mouse, Rabbit, Rat
Catalog No. 50-288-343